2-Bromo-1-((2-(trimethylsilyl)ethoxy)methyl)-1H-imidazole-4-carbonitrile 95%
Supplier: Ambeed
|
Warning
|
A616940-100MG
A616940-1G
A616940-250MG
77104-196EA
116.7
USD
77104-196
77104-198
77104-200
2-Bromo-1-((2-(trimethylsilyl)ethoxy)methyl)-1H-imidazole-4-carbonitrile 95%
2-Bromo-1-((2-(trimethylsilyl)ethoxy)methyl)-1H-imidazole-4-carbonitrile
Precaución: for Research Use Only
Formula:
C₁₀H₁₆BrN₃OSi MW: 302.24 g/mol |
MDL Number:
MFCD12964153 CAS Number: 854044-51-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...