2-(3,5-Dimethylphenyl)piperidine 95%

Supplier: Ambeed
Warning

A615393-1G A615393-250MG
77104-192EA 326.76 USD
77104-192 77104-194
2-(3,5-Dimethylphenyl)piperidine 95%
2-(3,5-Dimethylphenyl)piperidine

Precaución: for Research Use Only

Formula: C₁₃H₁₉N
MW: 189.297 g/mol
MDL Number: MFCD02663617
CAS Number: 383128-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR