4-(2-Methoxyphenyl)-3-oxobutyric acid methyl ester 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-(2-methoxyphenyl)-3-oxobutanoate

A615276-100MG A615276-1G A615276-250MG
77104-186EA 41.38 USD
77104-186 77104-188 77104-190
4-(2-Methoxyphenyl)-3-oxobutyric acid methyl ester 95%
4-(2-Methoxyphenyl)-3-oxobutyric acid methyl ester

Precaución: for Research Use Only

Formula: C₁₂H₁₄O₄
MW: 222.24 g/mol
MDL Number: MFCD08457133
CAS Number: 131644-96-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR