8-Chloro-5,10-dihydro-11H-dibenzo[b,e][1,4]-diazepin-11-one 98%

Supplier: Ambeed
Warning

A615273-1G A615273-5G
77104-182EA 55.17 USD
77104-182 77104-184
8-Chloro-5,10-dihydro-11H-dibenzo[b,e][1,4]-diazepin-11-one 98%
8-Chloro-5,10-dihydro-11H-dibenzo[b,e][1,4]-diazepin-11-one

Precaución: for Research Use Only

Formula: C₁₃H₉ClN₂O
MW: 244.67 g/mol
MDL Number: MFCD01863368
CAS Number: 50892-62-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR