(2,4-Dichlorobenzyl)methylamine 95%

Supplier: Ambeed
Danger

Synonyms: 2,4-Dichloro-N-methylbenzylamine, N-Methyl-2,4-dichlorobenzylamine

A613548-1G A613548-25G A613548-5G
77104-176EA 11.67 USD
77104-176 77104-178 77104-180
(2,4-Dichlorobenzyl)methylamine 95%
(2,4-Dichlorobenzyl)methylamine

Precaución: for Research Use Only

Formula: C₈H₉Cl₂N
MW: 190.07 g/mol
MDL Number: MFCD00045185
CAS Number: 5013-77-4
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR