6-(3,5-Difluorophenyl)picolinic acid 97%

Supplier: Ambeed
Warning

A612394-100MG A612394-1G
77104-170EA 186.72 USD
77104-170 77104-172
6-(3,5-Difluorophenyl)picolinic acid 97%
6-(3,5-Difluorophenyl)picolinic acid

Precaución: for Research Use Only

Formula: C₁₂H₇F₂NO₂
MW: 235.19 g/mol
MDL Number: MFCD11847143
CAS Number: 1226098-50-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR