2,4-Dichloro-7-phenyl-6,7-dihydro-5H-cyclopenta[d]pyrimidine 95+%

Supplier: Ambeed
Warning

A612142-100MG A612142-250MG
77104-160EA 137.92 USD
77104-160 77104-162
2,4-Dichloro-7-phenyl-6,7-dihydro-5H-cyclopenta[d]pyrimidine 95+%
2,4-Dichloro-7-phenyl-6,7-dihydro-5H-cyclopenta[d]pyrimidine

Precaución: for Research Use Only

Formula: C₁₃H₁₀Cl₂N₂
MW: 265.13 g/mol
MDL Number: MFCD21607364
CAS Number: 1263868-24-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR