2-Amino-N,N-diethylacetamide 95%

Supplier: Ambeed
Warning

A612104-10G A612104-250MG
77104-156EA 678 USD
77104-156 77104-158
2-Amino-N,N-diethylacetamide 95%
2-Amino-N,N-diethylacetamide

Precaución: for Research Use Only

Formula: C₆H₁₄N₂O
MDL Number: MFCD08060128
CAS Number: 34105-57-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR