6-Chloro-1H-pyrrolo[3,2-c]pyridine-3-carbonitrile 95%

Supplier: Ambeed
Danger

A610365-100MG A610365-250MG
77104-134EA 42.44 USD
77104-134 77104-136
6-Chloro-1H-pyrrolo[3,2-c]pyridine-3-carbonitrile 95%
6-Chloro-1H-pyrrolo[3,2-c]pyridine-3-carbonitrile

Precaución: for Research Use Only

Formula: C₈H₄ClN₃
MW: 177.59 g/mol
MDL Number: MFCD18380880
CAS Number: 1260381-90-5
UN: 3439
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR