4-Bromo-1-fluoro-2-(trifluoromethoxy)benzene 95%

Supplier: Ambeed
Danger

Synonyms: 2-Fluoro-5-bromo-trifluoromethoxybenzene, 5-Bromo-2-fluorophenyl trifluoromethyl ether, 5-Bromo-α,α,α,2-tetrafluoroanisole

A609928-5G A609928-1G A609928-250MG A609928-25G
77104-122EA 110 USD
77104-122 77901-634 77900-903 77901-065
4-Bromo-1-fluoro-2-(trifluoromethoxy)benzene 95%
4-Bromo-1-fluoro-2-(trifluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₇H₃BrF₄O
MDL Number: MFCD06660160
CAS Number: 886496-45-3
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR