Ethyl 7-bromo-2,2-dimethylheptanoate 97%

Supplier: Ambeed
Danger

A606282-1G A606282-25G A606282-5G
77104-114EA 6.37 USD
77104-114 77104-116 77104-118
Ethyl 7-bromo-2,2-dimethylheptanoate 97%
Ethyl 7-bromo-2,2-dimethylheptanoate

Precaución: for Research Use Only

Formula: C₁₁H₂₁BrO₂
MW: 265.18 g/mol
MDL Number: MFCD27942427
CAS Number: 123469-92-1
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR