(2R,5R)-1,6-Diphenylhexane-2,5-diamine dihydrochloride 98%

Supplier: Ambeed
Warning

A604908-1G A604908-250MG A604908-5G
77104-108EA 29 USD
77104-108 77104-110 77104-112
(2R,5R)-1,6-Diphenylhexane-2,5-diamine dihydrochloride 98%
(2R,5R)-1,6-Diphenylhexane-2,5-diamine dihydrochloride

Precaución: for Research Use Only

Formula: C₁₈H₂₆Cl₂N₂
MDL Number: MFCD28100861
CAS Number: 1247119-31-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR