(Bis(trifluoroacetoxy)iodo)benzene 97%

Supplier: Ambeed
Warning

Synonyms: [Bis(trifluoroacetoxy)](phenyl)iodane

A604530-100G A604530-10G A604530-25G A604530-5G
77104-100EA 35 USD
77104-100 77104-102 77104-104 77104-106
(Bis(trifluoroacetoxy)iodo)benzene 97%
(Bis(trifluoroacetoxy)iodo)benzene

Precaución: for Research Use Only

Formula: C₁₀H₅F₆IO₄
MDL Number: MFCD00009672
CAS Number: 2712-78-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR