3-((4-Aminophenyl)sulfonyl)propan-1-ol 95+%
Supplier: Ambeed
|
Warning
|
A603898-1G
A603898-250MG
A603898-5G
77104-088EA
64
USD
77104-088
77104-090
77104-092
3-((4-Aminophenyl)sulfonyl)propan-1-ol 95+%
3-((4-Aminophenyl)sulfonyl)propan-1-ol
Precaución: for Research Use Only
Formula:
C₉H₁₃NO₃S MW: 215.26 g/mol |
MDL Number:
MFCD24643666 CAS Number: 131110-21-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...