[4-(Ethanesulfonyl)phenyl]methanamine hydrochloride 95%

Supplier: Ambeed
Warning

A603648-100MG A603648-1G A603648-250MG
77104-076EA 17 USD
77104-076 77104-078 77104-080
[4-(Ethanesulfonyl)phenyl]methanamine hydrochloride 95%
[4-(Ethanesulfonyl)phenyl]methanamine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₄ClNO₂S
MDL Number: MFCD28118402
CAS Number: 98959-89-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR