Fmoc-Lys(Pal-Glu-OtBu)-OH 98%

Supplier: Ambeed
Warning

Synonyms: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-6-((S)-5-(tert-butoxy)-5-oxo-4-palmitamidopentanamido)hexanoic acid

A603621-1G A603621-250MG A603621-5G
77104-072EA 24 USD
77104-072 77104-074 77551-814
Fmoc-Lys(Pal-Glu-OtBu)-OH 98%
Fmoc-Lys(Pal-Glu-OtBu)-OH

Precaución: for Research Use Only

Formula: C₄₆H₆₉N₃O₈
MDL Number: MFCD27952849
CAS Number: 1491158-62-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR