1-((2-(Trimethylsilyl)ethoxy)methyl)-1H-pyrazol-3-amine 95%

Supplier: Ambeed
Warning

A601598-100MG A601598-1G A601598-250MG
77104-050EA 130.5 USD
77104-050 77104-052 77104-054
1-((2-(Trimethylsilyl)ethoxy)methyl)-1H-pyrazol-3-amine 95%
1-((2-(Trimethylsilyl)ethoxy)methyl)-1H-pyrazol-3-amine

Precaución: for Research Use Only

Formula: C₉H₁₉N₃OSi
MW: 213.35 g/mol
MDL Number: MFCD24623294
CAS Number: 885325-91-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR