5-(2-Hydroxy-2-propyl)-2-methylphenylboronic acid pinacol ester 95%

Supplier: Ambeed
Warning

A601551-100MG A601551-1G A601551-250MG
77104-044EA 42.44 USD
77104-044 77104-046 77104-048
5-(2-Hydroxy-2-propyl)-2-methylphenylboronic acid pinacol ester 95%
5-(2-Hydroxy-2-propyl)-2-methylphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₆H₂₅BO₃
MW: 276.179 g/mol
MDL Number: MFCD18730087
CAS Number: 1345961-02-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR