tert-Butyl 2-(4-bromophenyl)propan-2-ylcarbamate 95%

Supplier: Ambeed
Warning

A599002-100MG A599002-250MG A599002-5G
77104-004EA 22 USD
77104-004 77104-006 77104-008
tert-Butyl 2-(4-bromophenyl)propan-2-ylcarbamate 95%
tert-Butyl 2-(4-bromophenyl)propan-2-ylcarbamate

Precaución: for Research Use Only

Formula: C₁₄H₂₀BrNO₂
MDL Number: MFCD16619489
CAS Number: 214973-83-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR