3-Propionylpyridine 98%

Supplier: Ambeed
Warning

Synonyms: 1-(Pyridin-3-yl)propan-1-one

A598396-1G A598396-25G A598396-5G
77103-998EA 32.89 USD
77103-998 77104-000 77104-002
3-Propionylpyridine 98%
3-Propionylpyridine

Precaución: for Research Use Only

Formula: C₈H₉NO
MW: 135.17 g/mol
MDL Number: MFCD00023589
CAS Number: 1570-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR