2-Iodoterephthalic acid 98%

Supplier: Ambeed
Warning

A597630-100MG A597630-1G A597630-250MG A597630-5G
77103-990EA 20 USD
77103-990 77103-992 77103-994 77103-996
2-Iodoterephthalic acid 98%
2-Iodoterephthalic acid

Precaución: for Research Use Only

Formula: C₈H₅IO₄
MDL Number: MFCD00962043
CAS Number: 1829-22-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR