3-(Aminomethyl)-4-methoxy-6-methylpyridin-2-ol 95%

Supplier: Ambeed
Warning

A596008-100MG A596008-1G A596008-250MG A596008-5G
77103-980EA 82.76 USD
77103-980 77103-982 77103-984 77103-986
3-(Aminomethyl)-4-methoxy-6-methylpyridin-2-ol 95%
3-(Aminomethyl)-4-methoxy-6-methylpyridin-2-ol

Precaución: for Research Use Only

Formula: C₈H₁₂N₂O₂
MW: 168.19 g/mol
MDL Number: MFCD27952025
CAS Number: 1438382-15-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR