2,4-Difluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline 95%

Supplier: Ambeed
Warning

A594452-100MG A594452-1G A594452-250MG
77103-966EA 37.14 USD
77103-966 77103-968 77103-970
2,4-Difluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline 95%
2,4-Difluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

Precaución: for Research Use Only

Formula: C₁₂H₁₆BF₂NO₂
MW: 255.06 g/mol
MDL Number: MFCD28291811
CAS Number: 1415568-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR