3-Bromo-5-(1-hydroxyethyl)pyridine 98%

Supplier: Ambeed
Warning

A594006-1G A594006-250MG
77103-964EA 45 USD
77103-964 77901-449
3-Bromo-5-(1-hydroxyethyl)pyridine 98%
3-Bromo-5-(1-hydroxyethyl)pyridine

Precaución: for Research Use Only

Formula: C₇H₈BrNO
MDL Number: MFCD13188109
CAS Number: 283608-66-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR