Ethyl 4-amino-4-oxobutanoate 98%

Supplier: Ambeed
Warning

A593950-100G A593950-1G A593950-25G A593950-5G
77103-956EA 690 USD
77103-956 77103-958 77103-960 77103-962
Ethyl 4-amino-4-oxobutanoate 98%
Ethyl 4-amino-4-oxobutanoate

Precaución: for Research Use Only

Formula: C₆H₁₁NO₃
MDL Number: MFCD03839938
CAS Number: 53171-35-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR