N-((1R,2R)-2-Amino-1,2-diphenylethyl)-2,4,6-trimethylbenzenesulfonamide ≥97%

Supplier: Ambeed
Warning

A593777-100MG A593777-1G A593777-250MG
77103-950EA 13 USD
77103-950 77103-952 77103-954
N-((1R,2R)-2-Amino-1,2-diphenylethyl)-2,4,6-trimethylbenzenesulfonamide ≥97%
N-((1R,2R)-2-Amino-1,2-diphenylethyl)-2,4,6-trimethylbenzenesulfonamide

Precaución: for Research Use Only

Formula: C₂₃H₂₆N₂O₂S
MDL Number: MFCD11501122
CAS Number: 852212-90-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR