4-(Methylsulphonyl)benzyl bromide 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
α-Bromo-4-(methylsulphonyl)toluene, 1-(Bromomethyl)-4-(methylsulphonyl)benzene
A592991-100G
A592991-1G
A592991-250MG
A592991-5G
A592991-10G
A592991-25G
77103-936EA
494
USD
77103-936
77103-938
77103-940
77103-942
77686-504
77686-505
4-(Methylsulphonyl)benzyl bromide 98%
4-(Methylsulphonyl)benzyl bromide
Precaución: for Research Use Only
Formula:
C₈H₉BrO₂S |
MDL Number:
MFCD00185849 CAS Number: 53606-06-7 UN: 3261 ADR: 8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...