2-(5-Ethoxy-2,3-difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
Supplier: Ambeed
|
Warning
|
A592337-100MG
A592337-1G
A592337-250MG
77103-930EA
36.08
USD
77103-930
77103-932
77103-934
2-(5-Ethoxy-2,3-difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
2-(5-Ethoxy-2,3-difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Precaución: for Research Use Only
Formula:
C₁₄H₁₉BF₂O₃ MW: 284.10 g/mol |
MDL Number:
MFCD19237209 CAS Number: 1445601-61-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...