4-Bromo-2-fluoroaniline hydrochloride 95+%

Supplier: Ambeed
Warning

A592281-100MG A592281-1G A592281-250MG
77103-924EA 22 USD
77103-924 77103-926 77103-928
4-Bromo-2-fluoroaniline hydrochloride 95+%
4-Bromo-2-fluoroaniline hydrochloride

Precaución: for Research Use Only

Formula: C₆H₆BrClFN
MDL Number: MFCD07778061
CAS Number: 1174029-29-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR