1-Methyl-3-octylimidazolium bromide 96%

Supplier: Ambeed
Warning

A591908-100G A591908-5G
77103-920EA 56 USD
77103-920 77103-922
1-Methyl-3-octylimidazolium bromide 96%
1-Methyl-3-octylimidazolium bromide

Precaución: for Research Use Only

Formula: C₁₂H₂₃BrN₂
MDL Number: MFCD10566918
CAS Number: 61545-99-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR