LH-846 98%

Supplier: Ambeed
Warning

Synonyms: LH846, N-(5-Chloro-6-methylbenzo[d]thiazol-2-yl)-2-phenylacetamide

A591543-10MG A591543-1MG A591543-25MG A591543-50MG A591543-5MG
77103-910EA 41.38 USD
77103-910 77103-912 77103-914 77103-916 77103-918
LH-846 98%
LH-846

Precaución: for Research Use Only

Formula: C₁₆H₁₃ClN₂OS
MW: 316.81 g/mol
MDL Number: MFCD03786038
CAS Number: 639052-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR