1-(2-Ethynylphenyl)ethanone 95%

Supplier: Ambeed
Warning

A589470-1G A589470-250MG
77103-884EA 422.24 USD
77103-884 77103-886
1-(2-Ethynylphenyl)ethanone 95%
1-(2-Ethynylphenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₀H₈O
MW: 144.16 g/mol
MDL Number: MFCD18207358
CAS Number: 104190-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR