6-Chloro-5-cyanonicotinic acid 95%

Supplier: Ambeed
Warning

A588589-100MG A588589-250MG
77103-880EA 29 USD
77103-880 77103-882
6-Chloro-5-cyanonicotinic acid 95%
6-Chloro-5-cyanonicotinic acid

Precaución: for Research Use Only

Formula: C₇H₃ClN₂O₂
MDL Number: MFCD14585457
CAS Number: 1206969-20-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR