(R)-4-Methyl-2-(hydroxymethyl)morpholine 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
(R)-(4-Methylmorpholin-2-yl)methanol
A588552-100MG
A588552-250MG
A588552-1G
A588552-5G
77103-876EA
41.38
USD
77103-876
77103-878
77594-788
77613-828
(R)-4-Methyl-2-(hydroxymethyl)morpholine 95%
(R)-4-Methyl-2-(hydroxymethyl)morpholine
Precaución: for Research Use Only
Formula:
C₆H₁₃NO₂ MW: 131.173 g/mol |
MDL Number:
MFCD16294696 CAS Number: 1159598-35-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...