1-Bromo-3-fluoro-2-methyl-5-nitrobenzene 95%

Supplier: Ambeed
Warning

A587520-1G A587520-250MG
77103-862EA 125.19 USD
77103-862 77103-864
1-Bromo-3-fluoro-2-methyl-5-nitrobenzene 95%
1-Bromo-3-fluoro-2-methyl-5-nitrobenzene

Precaución: for Research Use Only

Formula: C₇H₅BrFNO₂
MW: 234.02 g/mol
MDL Number: MFCD04973754
CAS Number: 207110-34-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR