6'-Bromospiro[cyclopropane-1,3'-indolin]-2'-one 95%
Supplier: Ambeed
|
Warning
|
A587154-250MG
A587154-1G
77103-860EA
231.28
USD
77103-860
77686-489
6'-Bromospiro[cyclopropane-1,3'-indolin]-2'-one 95%
6'-Bromospiro[cyclopropane-1,3'-indolin]-2'-one
Precaución: for Research Use Only
Formula:
C₁₀H₈BrNO MW: 238.08 g/mol |
MDL Number:
MFCD19703307 CAS Number: 1378865-57-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...