(2S,4S)-4-Fluoropyrrolidine-2-carbonitrile hydrochloride 98%

Supplier: Ambeed
Warning

A586564-100MG A586564-1G A586564-250MG A586564-5G
77103-852EA 44 USD
77103-852 77103-854 77103-856 77103-858
(2S,4S)-4-Fluoropyrrolidine-2-carbonitrile hydrochloride 98%
(2S,4S)-4-Fluoropyrrolidine-2-carbonitrile hydrochloride
Formula: C₅H₈ClFN₂
MDL Number: MFCD28014566
CAS Number: 426844-77-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR