3,4-Dichlorobenzyl alcohol 98%

Supplier: Ambeed
Warning

Synonyms: (3,4-Dichlorophenyl)methanol

A585955-10G A585955-25G
77103-838EA 11 USD
77103-838 77103-840 77103-842
3,4-Dichlorobenzyl alcohol 98%
3,4-Dichlorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₆Cl₂O
MDL Number: MFCD00004633
CAS Number: 1805-32-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR