4-Fluoro-2-iodoaniline 96%

Supplier: Ambeed
Warning

A584609-100G A584609-10G A584609-25G A584609-5G
77103-826EA 77 USD
77103-826 77103-828 77103-830 77103-832
4-Fluoro-2-iodoaniline 96%
4-Fluoro-2-iodoaniline

Precaución: for Research Use Only

Formula: C₆H₅FIN
MDL Number: MFCD00042584
CAS Number: 61272-76-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR