3-Chloro-4-fluoroaniline 98%

Supplier: Ambeed
Warning

A584254-1000G A584254-100G A584254-25G A584254-500G
77103-818EA 61.54 USD
77103-818 77103-820 77103-822 77103-824
3-Chloro-4-fluoroaniline 98%
3-Chloro-4-fluoroaniline

Precaución: for Research Use Only

Formula: C₆H₅ClFN
MW: 145.56 g/mol
MDL Number: MFCD00007767
CAS Number: 367-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR