3,6-Bis(5-bromo-2-thienyl)-2,5-bis(2-hexyldecyl)pyrrolo[3,4-c]pyrrole-1,4(2H,5H)-dione 95%

Supplier: Ambeed
Warning

A581634-100MG A581634-1G A581634-250MG
77103-782EA 19.1 USD
77103-782 77103-784 77103-786
3,6-Bis(5-bromo-2-thienyl)-2,5-bis(2-hexyldecyl)pyrrolo[3,4-c]pyrrole-1,4(2H,5H)-dione 95%
3,6-Bis(5-bromo-2-thienyl)-2,5-bis(2-hexyldecyl)pyrrolo[3,4-c]pyrrole-1,4(2H,5H)-dione

Precaución: for Research Use Only

Formula: C₄₆H₇₀Br₂N₂O₂S₂
MW: 906.99 g/mol
MDL Number: MFCD28098664
CAS Number: 1000623-98-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR