1,5-Dichloro-2-iodo-3-methoxybenzene 95%

Supplier: Ambeed
Warning

A580132-100MG A580132-250MG
77103-754EA 41.38 USD
77103-754 77103-756
1,5-Dichloro-2-iodo-3-methoxybenzene 95%
1,5-Dichloro-2-iodo-3-methoxybenzene

Precaución: for Research Use Only

Formula: C₇H₅Cl₂IO
MW: 302.92 g/mol
MDL Number: MFCD21607156
CAS Number: 1271523-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR