3-Acetyl-1H-pyrazole-5-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 5-Acetyl-1H-pyrazole-3-carboxylic acid

A579679-1G A579679-5G
77103-750EA 15.92 USD
77103-750 77103-752
3-Acetyl-1H-pyrazole-5-carboxylic acid 97%
3-Acetyl-1H-pyrazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₆N₂O₃
MW: 154.13 g/mol
MDL Number: MFCD05864511
CAS Number: 1297537-45-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR