1-Fluoro-3-Methoxy-2-methylbenzene 95%

Supplier: Ambeed
Warning

A577524-100MG A577524-1G A577524-250MG
77103-712EA 53.05 USD
77103-712 77103-714 77103-716
1-Fluoro-3-Methoxy-2-methylbenzene 95%
1-Fluoro-3-Methoxy-2-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₉FO
MW: 140.15 g/mol
MDL Number: MFCD12922638
CAS Number: 1159883-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR