4-(2-Aminoethyl)piperidine, 4-BOC protected 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-(2-Boc-aminoethyl)piperidine, tert-Butyl [2-(piperidin-4-yl)ethyl]carbamate, 4-[2-(Boc-amino)ethyl]piperidine, N-(tert-Butoxycarbonyl)-2-(piperidin-4-yl)ethylamine
A576385-1G
A576385-250MG
A576385-5G
A576385-25G
A576385-100MG
77103-706EA
36
USD
77103-706
77103-708
77103-710
77686-466
77900-782
4-(2-Aminoethyl)piperidine, 4-BOC protected 95%
4-(2-Aminoethyl)piperidine, 4-BOC protected
Precaución: for Research Use Only
Formula:
C₁₂H₂₄N₂O₂ MW: 228.331 g/mol |
MDL Number:
MFCD01318373 CAS Number: 165528-81-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...