Boc-Nα-methyl-4-chloro-L-phenylalanine 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-2-((tert-Butoxycarbonyl)(methyl)amino)-3-(4-chlorophenyl)propanoic acid, Boc-N-Me-Phe(4-Cl)-OH
A575638-100MG
A575638-250MG
77103-702EA
50
USD
77103-702
77103-704
Boc-Nα-methyl-4-chloro-L-phenylalanine 95%
Boc-Nalpha-methyl-4-chloro-L-phenylalanine
Precaución: for Research Use Only
Formula:
C₁₅H₂₀ClNO₄ MW: 313.78 g/mol |
MDL Number:
MFCD00672517 CAS Number: 179033-68-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...