2-Nitro-4-[(trifluoromethyl)thio]phenol 97%

Supplier: Ambeed
Warning

A574556-5G A574556-1G A574556-250MG
77103-698EA 42 USD
77103-698 77900-557 77901-126
2-Nitro-4-[(trifluoromethyl)thio]phenol 97%
2-Nitro-4-[(trifluoromethyl)thio]phenol

Precaución: for Research Use Only

Formula: C₇H₄F₃NO₃S
MDL Number: MFCD08457463
CAS Number: 933673-33-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR