Methyl 2-(4-chlorophenyl)-2-oxoacetate 95%

Supplier: Ambeed
Warning

A573344-1G A573344-250MG A573344-5G
77103-694EA 19 USD
77103-694 77103-696 77572-824
Methyl 2-(4-chlorophenyl)-2-oxoacetate 95%
Methyl 2-(4-chlorophenyl)-2-oxoacetate

Precaución: for Research Use Only

Formula: C₉H₇ClO₃
MDL Number: MFCD00157150
CAS Number: 37542-28-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR