4-Chloro-1-(2,4-difluorophenyl)butan-1-one 95%

Supplier: Ambeed
Warning

A572970-100MG A572970-1G A572970-250MG A572970-5G
77103-686EA 46.68 USD
77103-686 77103-688 77103-690 77103-692
4-Chloro-1-(2,4-difluorophenyl)butan-1-one 95%
4-Chloro-1-(2,4-difluorophenyl)butan-1-one

Precaución: for Research Use Only

Formula: C₁₀H₉ClF₂O
MW: 218.62 g/mol
MDL Number: MFCD21677506
CAS Number: 50528-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR