Z-Leu-OSu 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Z-Leu-Osu, N-Benzyloxycarbonyl-L-leucine N-succinimidyl ester
A568398-1G
A568398-5G
A568398-25G
77103-668EA
9.55
USD
77103-668
77613-774
77686-450
Z-Leu-OSu 95%
Z-Leu-OSu
Precaución: for Research Use Only
Formula:
C₁₈H₂₂N₂O₆ MW: 362.382 g/mol |
MDL Number:
MFCD00042759 CAS Number: 3397-35-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...